Assay Details
Target Gene Details
Entrez Gene ID: | 83770 |
Gene Name: | taste receptor, type 1, member 2 |
Gene Aliases: |
Gpr71, T1r2, TR2 |
Location: |
Chr.4:139653538-139670280 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas1r2 | NM_031873.1 | 1 | 69 | NP_114079.1 |
| AF337041.1 | 1 | 69 | AAK39438.1 | |
| AY032623.1 | 1 | 38 | AAK51604.1 | |
| BC137740.1 | 1 | 58 | ||
| BC144879.1 | 1 | 58 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map