Assay Details
Target Gene Details
Entrez Gene ID: | 109331 |
Gene Name: | ring finger protein 20 |
Gene Aliases: |
4833430L21Rik, AW540162, C79397, mKIAA4116 |
Location: |
Chr.4:49632011-49656886 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf20 | NM_001163263.1 | 16 | 2744 | NP_001156735.1 |
| NM_182999.2 | 17 | 2591 | NP_892044.1 | |
| AK220492.1 | 16 | 2660 | BAD90290.1 | |
| BC052482.1 | 17 | 2530 | AAH52482.1 | |
| BQ442966.1 | 4 | 506 | ||
| CN531555.1 |

Genomic Map