Assay Details
Target Gene Details
Entrez Gene ID: | 109331 |
Gene Name: | ring finger protein 20 |
Gene Aliases: |
4833430L21Rik, AW540162, C79397, mKIAA4116 |
Location: |
Chr.4:49632011-49656886 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf20 | NM_001163263.1 | NP_001156735.1 | ||
| NM_182999.2 | NP_892044.1 | |||
| AK048862.1 | BAE43337.1 | |||
| AK051278.1 | BAC34590.2 | |||
| AK053783.1 | BAC35521.1 | |||
| AK076501.1 | BAC36367.1 | |||
| AK082529.1 | ||||
| AK139883.1 | BAE24170.1 | |||
| AK163905.1 | ||||
| AK220492.1 | BAD90290.1 | |||
| BC052482.1 | AAH52482.1 |

Genomic Map