Assay Details
Target Gene Details
Entrez Gene ID: | 381605 |
Gene Name: | TBC1 domain family, member 2 |
Gene Aliases: |
A630005A06Rik, BC022630, Gm1037, PARIS-1, PARIS1 |
Location: |
Chr.4:46604390-46650877 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d2 | NM_198664.3 | NP_941066.3 | ||
| AK041359.1 | BAC30918.1 | |||
| AK148254.1 | BAE28440.1 | |||
| AK148289.1 |
More Information
Set Membership: |
|

Genomic Map