Assay Details
Target Gene Details
Entrez Gene ID: | 242646 |
Gene Name: | Tctex1 domain containing 4 |
Gene Aliases: |
4833401D15Rik |
Location: |
Chr.4:117124524-117128946 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tctex1d4 | NM_175030.2 | NP_778195.1 | ||
| AK029345.1 | BAC26409.1 | |||
| BC115474.2 |
More Information
Set Membership: |
|

Genomic Map