Assay Details
Target Gene Details
Entrez Gene ID: | 319885 |
Gene Name: | zinc finger, CCHC domain containing 7 |
Gene Aliases: |
4930572I07Rik, D4Wsu132e |
Location: |
Chr.4:44756559-44932215 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zcchc7 | NM_138590.4 | NP_613056.2 | ||
AK029808.1 | ||||
AK032353.1 | ||||
AK039158.1 | ||||
AK043238.1 | BAC31499.1 | |||
AK080785.1 | ||||
AK157728.1 | BAE34173.1 | |||
BC117020.1 | ||||
BC117022.1 | ||||
BC144774.1 | ||||
CF732773.1 |
