Assay Details
Target Gene Details
Entrez Gene ID: | 319885 |
Gene Name: | zinc finger, CCHC domain containing 7 |
Gene Aliases: |
4930572I07Rik, D4Wsu132e |
Location: |
Chr.4:44756559-44932215 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zcchc7 | NM_138590.4 | NP_613056.2 | ||
AK013175.1 | BAB28692.1 | |||
AK050430.1 | BAC34252.1 | |||
BC054405.1 | AAH54405.1 |
