Assay Details
Target Gene Details
Entrez Gene ID: | 68948 |
Gene Name: | family with sequence similarity 216, member A |
Gene Aliases: |
1500011H22Rik |
Location: |
Chr.5:122364584-122371963 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam216a | NM_026883.3 | NP_081159.1 | ||
| AK005211.1 | BAB23885.1 | |||
| AK182306.1 | ||||
| BC019498.1 | AAH19498.1 | |||
| BC057658.1 | AAH57658.1 | |||
| BC061033.1 | AAH61033.1 |

Genomic Map