Assay Details
Target Gene Details
Entrez Gene ID: | 231821 |
Gene Name: | ArfGAP with dual PH domains 1 |
Gene Aliases: |
4930431P11Rik, Centa1, GC1L, p42IP4 |
Location: |
Chr.5:139271876-139325464 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adap1 | NM_172723.4 | 10 | 1093 | NP_766311.2 |
| AK076754.1 | 9 | 1079 | BAC36467.1 | |
| AK076768.1 | 10 | 850 | BAC36474.1 | |
| BC056934.1 | 9 | 819 | AAH56934.1 | |
| BC144874.1 | 9 | 835 |

Genomic Map