Assay Details
Target Gene Details
Entrez Gene ID: | 330050 |
Gene Name: | family with sequence similarity 185, member A |
Gene Aliases: |
4631428I10, AI847670 |
Location: |
Chr.5:21424861-21488271 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam185a | NM_177869.4 | NP_808537.2 | ||
| AK030922.1 | ||||
| BC055342.1 | AAH55342.1 |

Genomic Map