Assay Details
Target Gene Details
Entrez Gene ID: | 330188 |
Gene Name: | coiled-coil domain containing 63 |
Gene Aliases: |
4921511C16Rik |
Location: |
Chr.5:122108052-122138080 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc63 | NM_001289809.1 | NP_001276738.1 | ||
| NM_001289811.1 | NP_001276740.1 | |||
| NM_183307.4 | NP_899130.1 | |||
| AK029513.1 | BAC26487.1 | |||
| BC118944.1 | ||||
| BC118945.1 |
Target Gene Details
Entrez Gene ID: | 17906 |
Gene Name: | myosin, light polypeptide 2, regulatory, cardiac, slow |
Gene Aliases: |
MLC-2, MLC-2v, Mlc2v, Mylpc |
Location: |
Chr.5:122100904-122113396 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myl2 |
More Information
Set Membership: |
|

Genomic Map