Assay Details
Target Gene Details
Entrez Gene ID: | 330188 |
Gene Name: | coiled-coil domain containing 63 |
Gene Aliases: |
4921511C16Rik |
Location: |
Chr.5:122108052-122138080 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc63 | NM_001289809.1 | 12 | 1913 | NP_001276738.1 |
| NM_001289811.1 | 13 | 2005 | NP_001276740.1 | |
| NM_183307.4 | 13 | 2147 | NP_899130.1 | |
| AK029513.1 | 13 | 2149 | BAC26487.1 | |
| AV276792.1 | 1 | 79 | ||
| AV278877.1 | 2 | 135 | ||
| AV279232.1 | 1 | 115 | ||
| AV280582.1 | 1 | 100 | ||
| AV281915.1 | ||||
| BC118944.1 | 12 | 1835 | ||
| BC118945.1 | 13 | 1927 |
Target Gene Details
Entrez Gene ID: | 17906 |
Gene Name: | myosin, light polypeptide 2, regulatory, cardiac, slow |
Gene Aliases: |
MLC-2, MLC-2v, Mlc2v, Mylpc |
Location: |
Chr.5:122100904-122113396 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myl2 |

Genomic Map