Assay Details
Target Gene Details
Entrez Gene ID: | 13113 |
Gene Name: | cytochrome P450, family 3, subfamily a, polypeptide 13 |
Gene Aliases: |
IIIAm2 |
Location: |
Chr.5:137892933-137921619 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp3a13 | NM_007819.4 | NP_031845.1 | ||
| AK136494.1 | BAE23008.1 | |||
| AK136529.1 | BAE23029.1 | |||
| AK149572.1 | BAE28966.1 | |||
| BC046592.1 | AAH46592.1 | |||
| X63023.1 | CAA44754.1 |
More Information
Set Membership: |
|

Genomic Map