Assay Details
Target Gene Details
Entrez Gene ID: | 68184 |
Gene Name: | density-regulated protein |
Gene Aliases: |
1500003K04Rik |
Location: |
Chr.5:123907269-123928832 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Denr | NM_026603.4 | NP_080879.1 | ||
| AK010394.1 | ||||
| AK017647.1 | BAB30854.1 | |||
| AK018762.1 | BAB31392.1 | |||
| AK135127.1 | BAE22432.1 | |||
| BC043922.1 | AAH43922.1 |
More Information
Set Membership: |
|

Genomic Map