Assay Details
Target Gene Details
Entrez Gene ID: | 231821 |
Gene Name: | ArfGAP with dual PH domains 1 |
Gene Aliases: |
4930431P11Rik, Centa1, GC1L, p42IP4 |
Location: |
Chr.5:139271876-139325464 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adap1 | NM_172723.4 | NP_766311.2 | ||
| AK076754.1 | BAC36467.1 | |||
| AK076768.1 | BAC36474.1 | |||
| BC056934.1 | AAH56934.1 | |||
| BC144874.1 | ||||
| CX242691.1 |

Genomic Map