Assay Details
Target Gene Details
Entrez Gene ID: | 13114 |
Gene Name: | cytochrome P450, family 3, subfamily a, polypeptide 16 |
Gene Aliases: |
- |
Location: |
Chr.5:145436309-145469737 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp3a16 | NM_007820.2 | 13 | 1663 | NP_031846.2 |
| BC113143.1 | 13 | 1628 | AAI13144.1 | |
| D26137.1 | 13 | 1634 | BAA05133.1 |
More Information
Set Membership: |
|

Genomic Map