Assay Details
Target Gene Details
Entrez Gene ID: | 213948 |
Gene Name: | autophagy related 9B |
Gene Aliases: |
Apg912, Apg9l2, Apgdc2, Gm574, Nos3as, eONE, sONE |
Location: |
Chr.5:24384181-24392143 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atg9b | NM_001002897.3 | NP_001002897.2 | ||
| AY495541.1 | AAS72555.1 | |||
| AY515313.1 | AAS87214.1 |

Genomic Map