Assay Details
Target Gene Details
Entrez Gene ID: | 12453 |
Gene Name: | cyclin I |
Gene Aliases: |
- |
Location: |
Chr.5:93181933-93206495 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccni | NM_017367.3 | NP_059063.2 | ||
| AF005886.2 | AAD01253.2 | |||
| AK050465.1 | BAC34271.1 | |||
| AK079476.1 | ||||
| AK152614.1 | ||||
| AK208767.1 | ||||
| BC003290.1 | AAH03290.1 |
More Information
Set Membership: |
|

Genomic Map