Assay Details
Target Gene Details
Entrez Gene ID: | 70977 |
Gene Name: | calcium binding protein, spermatid specific 1 |
Gene Aliases: |
4931407G18Rik, AI428159, Clph |
Location: |
Chr.5:87979451-87981541 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cabs1 | NM_027631.2 | NP_081907.1 | ||
| AK076636.1 | BAC36426.1 | |||
| BC049750.1 | AAH49750.1 |
Target Gene Details
Entrez Gene ID: | 20599 |
Gene Name: | submaxillary gland androgen regulated protein 3A |
Gene Aliases: |
AA589601, MSG1, MSG3, Smr1, Smr3 |
Location: |
Chr.5:87969504-88008534 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smr3a |

Genomic Map