Assay Details
Target Gene Details
Entrez Gene ID: | 231045 |
Gene Name: | RIKEN cDNA 4931409K22 gene |
Gene Aliases: |
Iqca1l, Iqca1p1 |
Location: |
Chr.5:24543434-24555469 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931409K22Rik | NM_177676.6 | 15 | 1807 | NP_808344.3 |
| NR_033188.1 | 14 | 2297 | ||
| AK029839.1 | 14 | 2239 | BAC26640.1 | |
| BC137999.1 | 11 | 1528 | ||
| BC145792.1 | 11 | 1530 |

Genomic Map