Assay Details
Target Gene Details
Entrez Gene ID: | 231045 |
Gene Name: | RIKEN cDNA 4931409K22 gene |
Gene Aliases: |
Iqca1l, Iqca1p1 |
Location: |
Chr.5:24543434-24555469 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931409K22Rik | NM_177676.6 | 17 | 2166 | NP_808344.3 |
| NR_033188.1 | 16 | 2656 | ||
| AK029839.1 | 16 | 2598 | BAC26640.1 | |
| AV271477.2 | 1 | 112 | ||
| BC137999.1 | 13 | 1887 | ||
| BC145792.1 | 13 | 1889 |

Genomic Map