Assay Details
Target Gene Details
Entrez Gene ID: | 18671 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 1A |
Gene Aliases: |
Abcb4, Evi32, Mdr1a, Mdr3, P-gp, Pgp, Pgy-3, Pgy3, mdr-3 |
Location: |
Chr.5:8567091-8748575 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb1a | NM_011076.2 | 9 | 1026 | NP_035206.2 |
| AY864315.1 | 8 | 889 | AAW56448.1 | |
| M24417.1 | 3 | 375 | AAA03243.1 | |
| M30697.1 | 9 | 1040 | AAA39517.1 | |
| M33581.1 | 9 | 1025 | AAA39514.1 |
More Information
Set Membership: |
|

Genomic Map