Assay Details
Target Gene Details
Entrez Gene ID: | 18669 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 1B |
Gene Aliases: |
Abcb1, Mdr1, Mdr1b, Pgy-1, Pgy1, mdr |
Location: |
Chr.5:8798147-8866315 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb1b | NM_011075.2 | NP_035205.1 | ||
| AK030328.1 | ||||
| BC141363.1 | ||||
| BC150811.1 | ||||
| M14757.1 | AAA79005.1 |
More Information
Set Membership: |
|

Genomic Map