Assay Details
Target Gene Details
Entrez Gene ID: | 140904 |
Gene Name: | calneuron 1 |
Gene Aliases: |
9630012C17Rik, Cabp8, MNCb-0849 |
Location: |
Chr.5:130369356-130846325 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Caln1 | NM_181045.1 | 2 | 221 | NP_851388.1 |
| AF282251.1 | 2 | 203 | AAK15156.1 | |
| AK035865.1 | 2 | 190 | BAC29219.1 | |
| BY127416.1 | 2 | 222 |

Genomic Map