Assay Details
Target Gene Details
Entrez Gene ID: | 26893 |
Gene Name: | COP9 (constitutive photomorphogenic) homolog, subunit 6 (Arabidopsis thaliana) |
Gene Aliases: |
Sgn3, VIP/MOV34 |
Location: |
Chr.5:138161102-138163984 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cops6 | NM_012002.3 | 10 | 973 | NP_036132.1 |
| AF071315.1 | 10 | 1010 | AAC33902.1 | |
| AK138409.1 | 7 | 1314 | ||
| AK146410.1 | 10 | 984 | ||
| AK146775.1 | 10 | 986 | BAE27424.1 | |
| BC004664.1 | 10 | 972 | AAH04664.1 | |
| BC014286.1 | 10 | 977 | AAH14286.1 |
More Information
Set Membership: |
|

Genomic Map