Assay Details
Target Gene Details
Entrez Gene ID: | 26893 |
Gene Name: | COP9 (constitutive photomorphogenic) homolog, subunit 6 (Arabidopsis thaliana) |
Gene Aliases: |
Sgn3, VIP/MOV34 |
Location: |
Chr.5:138161102-138163984 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cops6 | NM_012002.3 | NP_036132.1 | ||
| AF071315.1 | AAC33902.1 | |||
| AK138409.1 | ||||
| AK146410.1 | ||||
| AK146775.1 | BAE27424.1 | |||
| BC004664.1 | AAH04664.1 | |||
| BC014286.1 | AAH14286.1 |
More Information
Set Membership: |
|

Genomic Map