Assay Details
Target Gene Details
Entrez Gene ID: | 18670 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 4 |
Gene Aliases: |
Mdr2, Pgy-2, Pgy2, mdr-2 |
Location: |
Chr.5:8893598-8959226 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb4 | NM_008830.2 | 21 | 2761 | NP_032856.2 |
| AK050345.1 | 20 | 2714 | ||
| BC150687.1 | 21 | 2631 | ||
| CJ071650.1 | 3 | 312 | ||
| J03398.1 | 21 | 2761 | AAA39516.1 |
More Information
Set Membership: |
|

Genomic Map