Assay Details
Target Gene Details
Entrez Gene ID: | 18670 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 4 |
Gene Aliases: |
Mdr2, Pgy-2, Pgy2, mdr-2 |
Location: |
Chr.5:8893598-8959226 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb4 | NM_008830.2 | 23 | 2993 | NP_032856.2 |
| AA239419.1 | 2 | 126 | ||
| BC150687.1 | 23 | 2863 | ||
| J03398.1 | 23 | 2993 | AAA39516.1 |
More Information
Set Membership: |
|

Genomic Map