Assay Details
Target Gene Details
Entrez Gene ID: | 18670 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 4 |
Gene Aliases: |
Mdr2, Pgy-2, Pgy2, mdr-2 |
Location: |
Chr.5:8893598-8959226 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb4 | NM_008830.2 | NP_032856.2 | ||
| AK050345.1 | ||||
| AK144024.1 | ||||
| AK165484.1 | BAE38214.1 | |||
| BC150687.1 | ||||
| J03398.1 | AAA39516.1 |
More Information
Set Membership: |
|

Genomic Map