Assay Details
Target Gene Details
Entrez Gene ID: | 13121 |
Gene Name: | cytochrome P450, family 51 |
Gene Aliases: |
AI426508, CYPLI, Cyp51a1, Ldm, P450-14DM, P450LI |
Location: |
Chr.5:4080674-4104697 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp51 | NM_020010.2 | NP_064394.2 | ||
| AF166266.1 | AAF73986.1 | |||
| AK028355.1 | BAC25900.1 | |||
| AK028815.1 | BAC26134.1 | |||
| AK031059.1 | BAC27231.1 | |||
| AK075764.1 | ||||
| AK076983.1 | BAC36548.1 | |||
| AK202785.1 | ||||
| AK208162.1 | ||||
| BC031813.1 | AAH31813.1 |
More Information
Set Membership: |
|

Genomic Map