Assay Details
Target Gene Details
Entrez Gene ID: | 242894 |
Gene Name: | ARP3 actin-related protein 3B |
Gene Aliases: |
9630005C02, ARP11, AW047569, Arp3b, Arp3beta |
Location: |
Chr.5:25759972-25850688 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actr3b | NM_001004365.1 | NP_001004365.1 | ||
| AK035798.1 | ||||
| BC082279.1 | AAH82279.1 |
More Information
Set Membership: |
|

Genomic Map