Assay Details
Target Gene Details
Entrez Gene ID: | 108723 |
Gene Name: | caspase recruitment domain family, member 11 |
Gene Aliases: |
0610008L17Rik, 2410011D02Rik, BIMP3, CARMA1 |
Location: |
Chr.5:140872999-141000596 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Card11 | NM_175362.2 | 3 | 407 | NP_780571.2 |
| AK002346.1 | 3 | 289 | ||
| AK131134.1 | 3 | 407 | BAD21384.1 | |
| AY135367.1 | 3 | 303 | AAN10150.1 | |
| BC096592.1 | 3 | 393 | AAH96592.1 |

Genomic Map