Assay Details
Target Gene Details
Entrez Gene ID: | 338350 |
Gene Name: | acyl-Coenzyme A dehydrogenase family, member 12 |
Gene Aliases: |
9330129D05Rik |
Location: |
Chr.5:121598277-121618938 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acad12 | NM_178799.3 | NP_848914.2 | ||
| AK033963.1 | BAC28530.1 | |||
| AK080793.1 | BAC38023.1 |
More Information
Set Membership: |
|

Genomic Map