Assay Details
Target Gene Details
Entrez Gene ID: | 13445 |
Gene Name: | CDK2 (cyclin-dependent kinase 2)-associated protein 1 |
Gene Aliases: |
Apc10, Cdkap1, DORC1, Doc1, ST19, doc-1, p12 |
Location: |
Chr.5:124345439-124354628 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk2ap1 | NM_013812.2 | NP_038840.2 | ||
| AF011644.1 | AAB65752.1 | |||
| AK004852.1 | BAB23616.1 | |||
| BC083075.1 | AAH83075.1 | |||
| BC103772.1 | AAI03773.1 | |||
| BC147052.1 | ||||
| BC147053.1 | ||||
| BE623061.1 | ||||
| M88755.1 |
More Information
Set Membership: |
|

Genomic Map