Assay Details
Target Gene Details
Entrez Gene ID: | 12861 |
Gene Name: | cytochrome c oxidase subunit VIa polypeptide 1 |
Gene Aliases: |
VIaL |
Location: |
Chr.5:115345654-115348955 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox6a1 | NM_007748.3 | NP_031774.1 | ||
| AK002402.1 | BAB22073.1 | |||
| AK019318.1 | BAB31662.2 | |||
| AK166496.1 | BAE38808.1 | |||
| BC003807.1 | AAH03807.1 | |||
| BC052816.1 | AAH52816.1 | |||
| L06465.1 | AAA53066.1 | |||
| U08440.1 | AAA17836.1 |
More Information
Set Membership: |
|

Genomic Map