Assay Details
Target Gene Details
Entrez Gene ID: | 231207 |
Gene Name: | cytoplasmic polyadenylation element binding protein 2 |
Gene Aliases: |
A630055H10Rik, Cpe-bp2 |
Location: |
Chr.5:43232254-43289724 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpeb2 | NM_001177379.1 | 1 | 1526 | NP_001170850.1 |
| NM_175937.3 | 1 | 1526 | NP_787951.2 | |
| AB100307.1 | 2 | 159 | BAC57076.1 | |
| AK042065.1 | 2 | 162 | ||
| AK164866.1 | 2 | 189 | ||
| BC107349.1 | 2 | 136 | AAI07350.1 | |
| BC107350.1 | 2 | 136 | AAI07351.1 |
Target Gene Details
Entrez Gene ID: | 665934 |
Gene Name: | predicted gene 7854 |
Gene Aliases: |
EG665934, ENSMUSG00000054483 |
Location: |
Chr.5:43151686-43235354 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm7854 | NR_028417.1 | |||
| AK041797.1 | BAC31070.1 | |||
| AK138349.1 |
More Information
Set Membership: |
|

Genomic Map