Assay Details
Target Gene Details
Entrez Gene ID: | 319251 |
Gene Name: | RIKEN cDNA 9630001P10 gene |
Gene Aliases: |
- |
Location: |
Chr.5:45639275-45645469 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9630001P10Rik | NR_102378.1 | |||
| AK044526.1 | ||||
| AK080411.1 | ||||
| BU610152.1 |
Target Gene Details
Entrez Gene ID: | 58227 |
Gene Name: | family with sequence similarity 184, member B |
Gene Aliases: |
9630031F12Rik, AB041544 |
Location: |
Chr.5:45529705-45639501 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam184b | NM_021416.3 | NP_067391.3 | ||
| AB257856.1 | 1 | 72 | ||
| BY794536.1 | 1 | 33 | ||
| CF539972.1 |

Genomic Map