Assay Details
Target Gene Details
Entrez Gene ID: | 319251 |
Gene Name: | RIKEN cDNA 9630001P10 gene |
Gene Aliases: |
- |
Location: |
Chr.5:45639275-45645469 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9630001P10Rik | NR_102378.1 | |||
| AK044526.1 | ||||
| AK080411.1 | ||||
| BU610152.1 |

Genomic Map