Assay Details
Target Gene Details
Entrez Gene ID: | 27405 |
Gene Name: | ATP-binding cassette, sub-family G (WHITE), member 3 |
Gene Aliases: |
Abcp2, Mxr2 |
Location: |
Chr.5:104935057-104982802 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcg3 | NM_030239.2 | 13 | 1738 | NP_084515.2 |
| AF324242.1 | 13 | 1690 | AAK14241.1 | |
| AK030831.1 | 13 | 1740 | ||
| AK051880.1 | 13 | 1704 | BAC34799.1 | |
| AK156626.1 | 13 | 1706 | ||
| BC138772.1 | 13 | 1719 | ||
| BC138773.1 | 13 | 1718 |
More Information
Set Membership: |
|

Genomic Map