Assay Details
Target Gene Details
Entrez Gene ID: | 11839 |
Gene Name: | amphiregulin |
Gene Aliases: |
AR, Mcub, Sdgf |
Location: |
Chr.5:91139615-91148432 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Areg | NM_009704.3 | 2 | 328 | NP_033834.1 |
| AK018590.1 | 2 | 344 | BAB31296.1 | |
| AK020273.1 | 2 | 344 | ||
| BC009138.1 | 2 | 328 | AAH09138.1 | |
| CT010320.1 | 2 | 129 | CAJ18528.1 | |
| D12648.1 | 2 | 155 | BAA02169.1 |
More Information
Set Membership: |
|

Genomic Map