Assay Details
Target Gene Details
Entrez Gene ID: | 11496 |
Gene Name: | a disintegrin and metallopeptidase domain 22 |
Gene Aliases: |
2900022I03Rik, AI854032, MDC2 |
Location: |
Chr.5:8072348-8368268 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam22 | NM_001007220.2 | NP_001007221.1 | ||
| NM_001007221.2 | NP_001007222.1 | |||
| NM_001098225.1 | NP_001091695.1 | |||
| AB009674.1 | BAA83382.1 | |||
| BC117707.1 | ||||
| HM004095.1 |
More Information
Set Membership: |
|

Genomic Map