Assay Details
Target Gene Details
Entrez Gene ID: | 56443 |
Gene Name: | actin related protein 2/3 complex, subunit 1A |
Gene Aliases: |
0610010H08Rik, 1110030K07Rik, 41kDa, AA407347, Sid32, Sid329 |
Location: |
Chr.5:145083869-145108756 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arpc1a | NM_019767.2 | NP_062741.1 | ||
| AB024984.1 | BAA84685.1 | |||
| AK002475.1 | ||||
| AK003986.1 | BAB23107.3 | |||
| AK076107.1 | BAC36187.1 | |||
| AK145663.1 | BAE26575.1 | |||
| BC001988.1 | AAH01988.1 |
More Information
Set Membership: |
|

Genomic Map