Assay Details
Target Gene Details
Entrez Gene ID: | 73471 |
Gene Name: | RIKEN cDNA 1700066N21 gene |
Gene Aliases: |
- |
Location: |
Chr.5:87908588-87979351 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700066N21Rik | NR_045924.1 | |||
| AK006908.1 |

Genomic Map