Assay Details
Target Gene Details
Entrez Gene ID: | 11496 |
Gene Name: | a disintegrin and metallopeptidase domain 22 |
Gene Aliases: |
2900022I03Rik, AI854032, MDC2 |
Location: |
Chr.5:8072348-8368268 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam22 | NM_001098225.1 | NP_001091695.1 | ||
| AB179842.1 | BAD72803.1 | |||
| AB179843.1 | BAD72804.1 | |||
| AB179844.1 | BAD72805.1 | |||
| AB179845.1 | BAD72806.1 | |||
| AB179846.1 | BAD72807.1 | |||
| AB179847.1 | BAD72808.1 | |||
| AB179848.1 | BAD72809.1 | |||
| AB179849.1 | BAD72810.1 | |||
| AB179850.1 | BAD72811.1 | |||
| AB179851.1 | BAD72812.1 | |||
| AB179852.1 | BAD72813.1 | |||
| AB179857.1 | BAD72818.1 | |||
| AB179858.1 | BAD72819.1 | |||
| AB179859.1 | BAD72820.1 | |||
| AB179860.1 | BAD72821.1 | |||
| AB179861.1 | BAD72822.1 | |||
| AK034528.1 | BAC28742.1 | |||
| HM004095.1 |
More Information
Set Membership: |
|

Genomic Map