Assay Details
Target Gene Details
Entrez Gene ID: | 70797 |
Gene Name: | ankyrin repeat and IBR domain containing 1 |
Gene Aliases: |
2310061P20Rik, 4631416I11Rik, AW494740, C80642, mKIAA1386 |
Location: |
Chr.5:3689999-3803124 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankib1 | NM_001003909.4 | 8 | 1777 | NP_001003909.2 |
| NM_001289527.1 | 8 | 1777 | NP_001276456.1 | |
| NM_001289528.1 | 7 | 1659 | NP_001276457.1 | |
| NM_001289529.1 | 7 | 1249 | NP_001276458.1 | |
| AK129343.1 | 8 | 1578 | BAC98153.1 | |
| BC145511.1 | 8 | 1528 | ||
| BC150792.1 | 8 | 1528 |
More Information
Set Membership: |
|

Genomic Map