Assay Details
Target Gene Details
Entrez Gene ID: | 13112 |
Gene Name: | cytochrome P450, family 3, subfamily a, polypeptide 11 |
Gene Aliases: |
AI256190, Cyp3a, IIIAm1, Pcn |
Location: |
Chr.5:145854607-145879854 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp3a11 | NM_007818.3 | NP_031844.1 | ||
| AK149433.1 | BAE28873.1 | |||
| BC010528.1 | AAH10528.1 | |||
| BC094062.1 | AAH94062.1 | |||
| X60452.1 | CAA42981.1 |
More Information
Set Membership: |
|

Genomic Map