Assay Details
Target Gene Details
Entrez Gene ID: | 242915 |
Gene Name: | GRB2 associated, regulator of MAPK1-like |
Gene Aliases: |
Fam59b, Gm444 |
Location: |
Chr.5:30105196-30118380 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gareml | NM_001167879.1 | NP_001161351.1 | ||
| BC060268.1 | AAH60268.1 | |||
| BC113782.1 | AAI13783.1 | |||
| BC113783.1 |

Genomic Map