Assay Details
Target Gene Details
Entrez Gene ID: | 14467 |
Gene Name: | glioblastoma amplified sequence |
Gene Aliases: |
AV006093, Nipsnap2 |
Location: |
Chr.5:129725044-129758327 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gbas | NM_008095.4 | 8 | 711 | NP_032121.3 |
| AJ001261.1 | 8 | 689 | CAA04635.1 | |
| AK170336.1 | 8 | 657 | BAE41726.1 | |
| AK183347.1 | ||||
| AK187878.1 | 1 | 40 | ||
| AK193098.1 | 1 | 35 | ||
| BC055893.1 | 8 | 660 | AAH55893.1 |

Genomic Map