Assay Details
Target Gene Details
Entrez Gene ID: | 625175 |
Gene Name: | RIKEN cDNA 1700028E10 gene |
Gene Aliases: |
A630054D14, Gm6561 |
Location: |
Chr.5:151368675-151414083 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700028E10Rik | NR_045700.1 | |||
| AK006454.1 |
Target Gene Details
Entrez Gene ID: | 243362 |
Gene Name: | StAR-related lipid transfer (START) domain containing 13 |
Gene Aliases: |
DLC2, GT650 |
Location: |
Chr.5:151037515-151428256 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stard13 |
More Information
Set Membership: |
|

Genomic Map