Assay Details
Target Gene Details
Entrez Gene ID: | 66587 |
Gene Name: | Fas-activated serine/threonine kinase |
Gene Aliases: |
0610011K02Rik, 610011K02Rik |
Location: |
Chr.5:24441039-24445282 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fastk | NM_023229.2 | NP_075718.2 | ||
| AF168682.1 | AAF89660.1 | |||
| AK002552.1 | ||||
| AK003940.1 | BAB23086.3 | |||
| AK038596.1 | BAC30060.1 | |||
| AK052667.1 | BAC35089.1 | |||
| AK139374.1 | ||||
| AK151859.1 | BAE30748.1 | |||
| AK152369.1 | BAE31159.1 | |||
| AK159751.1 | BAE35342.1 | |||
| AK167346.1 | BAE39445.1 | |||
| AK186668.1 | ||||
| AK187385.1 | ||||
| BC013547.1 | AAH13547.1 |

Genomic Map